RIE156330377 Rieke Metals, Inc.

ethyl 8-oxotridecanoate

  • MDL number MFCD01320346
  • Cas Number : 898776-48-2
  • Molecular Weight : 256.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7589F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7589F
Availability details for
Quantity
  • Availability
  • Add to Cart