RIE156330378 Rieke Metals, Inc.

Ethyl 4-oxoheptanoate

  • MDL number MFCD01320347
  • Cas Number : 14369-94-9
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7590B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7590B
Availability details for
Quantity
  • Availability
  • Add to Cart