RIE156330381 Rieke Metals, Inc.

ethyl 8-oxoundecanoate

  • MDL number MFCD01320351
  • Cas Number : 858794-06-6
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7590F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7590F
Availability details for
Quantity
  • Availability
  • Add to Cart