RIE156330390 Rieke Metals, Inc.

ethyl 2-(2-oxo-2-phenylethyl)benzoate

  • MDL number MFCD07700164
  • Cas Number : 898776-66-4
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7604

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7604
Availability details for
Quantity
  • Availability
  • Add to Cart