RIE156330391 Rieke Metals, Inc.

Ethyl 3-propionylbenzoate

  • MDL number MFCD02261278
  • Cas Number : 898776-68-6
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7606

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7606
Availability details for
Quantity
  • Availability
  • Add to Cart