RIE156330392 Rieke Metals, Inc.

ethyl 4-(3,5-dimethylphenyl)-2-methyl-4-oxobutyrate

  • MDL number MFCD02261285
  • Cas Number : 898776-70-0
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7615

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7615
Availability details for
Quantity
  • Availability
  • Add to Cart