RIE156330397 Rieke Metals, Inc.

ethyl 6-(3,4-dichlorophenyl)-6-oxohexanoate

  • MDL number MFCD02261292
  • Cas Number : 898776-76-6
  • Molecular Weight : 303.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7620D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7620D
Availability details for
Quantity
  • Availability
  • Add to Cart