RIE156330401 Rieke Metals, Inc.

ethyl 5-(3-bromo-4-methylphenyl)-5-oxovalerate

  • MDL number MFCD02261296
  • Cas Number : 898776-84-6
  • Molecular Weight : 313.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7621C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7621C
Availability details for
Quantity
  • Availability
  • Add to Cart