RIE156330402 Rieke Metals, Inc.

ethyl 6-(3-bromo-4-methylphenyl)-6-oxohexanoate

  • MDL number MFCD03844210
  • Cas Number : 898776-86-8
  • Molecular Weight : 327.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7621D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7621D
Availability details for
Quantity
  • Availability
  • Add to Cart