RIE156330403 Rieke Metals, Inc.

ethyl 7-(3-bromo-4-methylphenyl)-7-oxoheptanoate

  • MDL number MFCD03844211
  • Cas Number : 898776-88-0
  • Molecular Weight : 341.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7621E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7621E
Availability details for
Quantity
  • Availability
  • Add to Cart