RIE156330404 Rieke Metals, Inc.

ethyl 8-(3-bromo-4-methylphenyl)-8-oxooctanoate

  • MDL number MFCD03844212
  • Cas Number : 898776-90-4
  • Molecular Weight : 355.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7621F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7621F
Availability details for
Quantity
  • Availability
  • Add to Cart