RIE156330407 Rieke Metals, Inc.

ethyl 6-(4-bromo-2-methylphenyl)-6-oxohexanoate

  • Cas Number : 898776-96-0
  • Molecular Weight : 327.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7622D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7622D
Availability details for
Quantity
  • Availability
  • Add to Cart