RIE156330408 Rieke Metals, Inc.

ethyl 7-(4-bromo-2-methylphenyl)-7-oxoheptanoate

  • MDL number MFCD03844214
  • Cas Number : 898776-98-2
  • Molecular Weight : 341.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7622E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7622E
Availability details for
Quantity
  • Availability
  • Add to Cart