RIE156330409 Rieke Metals, Inc.

ethyl 8-(4-bromo-2-methylphenyl)-8-oxooctanoate

  • MDL number MFCD03844215
  • Cas Number : 898777-00-9
  • Molecular Weight : 355.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7622F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7622F
Availability details for
Quantity
  • Availability
  • Add to Cart