RIE156330411 Rieke Metals, Inc.

ethyl 5-(3-bromo-5-methylphenyl)-5-oxovalerate

  • MDL number MFCD02261301
  • Cas Number : 898777-06-5
  • Molecular Weight : 313.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7624C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7624C
Availability details for
Quantity
  • Availability
  • Add to Cart