RIE156330412 Rieke Metals, Inc.

Ethyl 6-methyl-5-oxoheptanoate

  • MDL number MFCD02261304
  • Cas Number : 24071-98-5
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7631C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7631C
Availability details for
Quantity
  • Availability
  • Add to Cart