RIE156330416 Rieke Metals, Inc.

Ethyl 4-(2-iodophenyl)-4-oxobutyrate

  • MDL number MFCD02261309
  • Cas Number : 263273-52-5
  • Molecular Weight : 332.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7633B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7633B
Availability details for
Quantity
  • Availability
  • Add to Cart