RIE156330421 Rieke Metals, Inc.

Ethyl 4-(3-iodophenyl)-4-oxobutyrate

  • MDL number MFCD02261314
  • Cas Number : 898777-24-7
  • Molecular Weight : 332.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7634B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7634B
Availability details for
Quantity
  • Availability
  • Add to Cart