RIE156330427 Rieke Metals, Inc.

Ethyl 5-(4-iodophenyl)-5-oxovalerate

  • MDL number MFCD02261320
  • Cas Number : 898777-42-9
  • Molecular Weight : 346.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7635C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7635C
Availability details for
Quantity
  • Availability
  • Add to Cart