RIE156330428 Rieke Metals, Inc.

ethyl 6-(4-iodophenyl)-6-oxohexanoate

  • MDL number MFCD02261321
  • Cas Number : 854658-72-3
  • Molecular Weight : 360.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7635D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7635D
Availability details for
Quantity
  • Availability
  • Add to Cart