RIE156330429 Rieke Metals, Inc.

ethyl 7-(4-iodophenyl)-7-oxoheptanoate

  • MDL number MFCD02261322
  • Cas Number : 898777-45-2
  • Molecular Weight : 374.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7635E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7635E
Availability details for
Quantity
  • Availability
  • Add to Cart