RIE156330434 Rieke Metals, Inc.

ethyl 8-(3-nitrophenyl)-8-oxooctanoate

  • MDL number MFCD02261333
  • Cas Number : 898777-57-6
  • Molecular Weight : 307.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7637F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7637F
Availability details for
Quantity
  • Availability
  • Add to Cart