RIE156330435 Rieke Metals, Inc.

ethyl 4-(4-nitrophenyl)-4-oxobutyrate

  • MDL number MFCD02261334
  • Cas Number : 15118-70-4
  • Molecular Weight : 251.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7638B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7638B
Availability details for
Quantity
  • Availability
  • Add to Cart