RIE156330436 Rieke Metals, Inc.

ethyl 5-(4-nitrophenyl)-5-oxovalerate

  • MDL number MFCD02261335
  • Cas Number : 898777-59-8
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7638C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7638C
Availability details for
Quantity
  • Availability
  • Add to Cart