RIE156330438 Rieke Metals, Inc.

ethyl 7-(4-nitrophenyl)-7-oxoheptanoate

  • MDL number MFCD02261337
  • Cas Number : 898777-63-4
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7638E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7638E
Availability details for
Quantity
  • Availability
  • Add to Cart