RIE156330440 Rieke Metals, Inc.

Ethyl 4-oxo-4-(2-trifluoromethylphenyl)butyrate

  • MDL number MFCD02261339
  • Cas Number : 898777-67-8
  • Molecular Weight : 274.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7639B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7639B
Availability details for
Quantity
  • Availability
  • Add to Cart