RIE156330443 Rieke Metals, Inc.

ethyl 8-oxo-8-(2-trifluoromethylphenyl)octanoate

  • MDL number MFCD02261343
  • Cas Number : 898777-73-6
  • Molecular Weight : 330.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7639F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7639F
Availability details for
Quantity
  • Availability
  • Add to Cart