RIE156330445 Rieke Metals, Inc.

Ethyl 5-oxo-5-(3-trifluoromethylphenyl)valerate

  • MDL number MFCD02261345
  • Cas Number : 898777-75-8
  • Molecular Weight : 288.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7640C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7640C
Availability details for
Quantity
  • Availability
  • Add to Cart