RIE156330446 Rieke Metals, Inc.

Ethyl 6-oxo-6-(3-trifluoromethylphenyl)hexanoate

  • MDL number MFCD02261346
  • Cas Number : 898777-77-0
  • Molecular Weight : 302.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7640D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7640D
Availability details for
Quantity
  • Availability
  • Add to Cart