RIE156330447 Rieke Metals, Inc.

ethyl 7-oxo-7-(3-trifluoromethylphenyl)heptanoate

  • MDL number MFCD02261347
  • Cas Number : 122115-59-7
  • Molecular Weight : 316.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7640E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7640E
Availability details for
Quantity
  • Availability
  • Add to Cart