RIE156330452 Rieke Metals, Inc.

ethyl 4-(2,3-dichlorophenyl)-4-oxobutyrate

  • MDL number MFCD02261354
  • Cas Number : 71450-93-6
  • Molecular Weight : 275.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7642B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7642B
Availability details for
Quantity
  • Availability
  • Add to Cart