RIE156330455 Rieke Metals, Inc.

ethyl 7-(2,3-dichlorophenyl)-7-oxoheptanoate

  • MDL number MFCD02261357
  • Cas Number : 898777-91-8
  • Molecular Weight : 317.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7642E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7642E
Availability details for
Quantity
  • Availability
  • Add to Cart