RIE156330459 Rieke Metals, Inc.

ethyl 6-(2,4-dichlorophenyl)-6-oxohexanoate

  • MDL number MFCD02261361
  • Cas Number : 898777-99-6
  • Molecular Weight : 303.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7643D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7643D
Availability details for
Quantity
  • Availability
  • Add to Cart