RIE156330461 Rieke Metals, Inc.

ethyl 8-(2,4-dichlorophenyl)-8-oxooctanoate

  • MDL number MFCD02261363
  • Cas Number : 898778-05-7
  • Molecular Weight : 331.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7643F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7643F
Availability details for
Quantity
  • Availability
  • Add to Cart