RIE156330463 Rieke Metals, Inc.

ethyl 5-(2,5-dichlorophenyl)-5-oxovalerate

  • MDL number MFCD02261365
  • Cas Number : 898778-11-5
  • Molecular Weight : 289.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7644C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7644C
Availability details for
Quantity
  • Availability
  • Add to Cart