RIE156330467 Rieke Metals, Inc.

ethyl 4-(4-ethylphenyl)-4-oxobutyrate

  • MDL number MFCD02261374
  • Cas Number : 57821-79-1
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7646B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7646B
Availability details for
Quantity
  • Availability
  • Add to Cart