RIE156330469 Rieke Metals, Inc.

ethyl 6-(4-ethylphenyl)-6-oxohexanoate

  • MDL number MFCD02261376
  • Cas Number : 898778-26-2
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7646D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7646D
Availability details for
Quantity
  • Availability
  • Add to Cart