RIE156330471 Rieke Metals, Inc.

ethyl 8-(4-ethylphenyl)-8-oxooctanoate

  • MDL number MFCD02261378
  • Cas Number : 898778-32-0
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7646F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7646F
Availability details for
Quantity
  • Availability
  • Add to Cart