RIE156330472 Rieke Metals, Inc.

ethyl 4-(4-isopropylphenyl)-4-oxobutyrate

  • MDL number MFCD02261384
  • Cas Number : 26803-60-1
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7648B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7648B
Availability details for
Quantity
  • Availability
  • Add to Cart