RIE156330473 Rieke Metals, Inc.

ethyl 5-(4-isopropylphenyl)-5-oxovalerate

  • MDL number MFCD02261385
  • Cas Number : 898778-35-3
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7648C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7648C
Availability details for
Quantity
  • Availability
  • Add to Cart