RIE156330474 Rieke Metals, Inc.

ethyl 6-(4-isopropylphenyl)-6-oxohexanoate

  • MDL number MFCD02261386
  • Cas Number : 898778-38-6
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7648D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7648D
Availability details for
Quantity
  • Availability
  • Add to Cart