RIE156330475 Rieke Metals, Inc.

Ethyl 7-(4-isopropylphenyl)-7-oxoheptanoate

  • MDL number MFCD02261387
  • Cas Number : 122115-50-8
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7648E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7648E
Availability details for
Quantity
  • Availability
  • Add to Cart