RIE156330476 Rieke Metals, Inc.

Ethyl 8-(4-isopropylphenyl)-8-oxooctanoate

  • MDL number MFCD02261388
  • Cas Number : 898778-41-1
  • Molecular Weight : 304.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7648F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7648F
Availability details for
Quantity
  • Availability
  • Add to Cart