RIE156330477 Rieke Metals, Inc.

ethyl 4-(4-t-butylphenyl)-4-oxobutyrate

  • MDL number MFCD02261394
  • Cas Number : 75237-09-1
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7650B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7650B
Availability details for
Quantity
  • Availability
  • Add to Cart