RIE156330478 Rieke Metals, Inc.

ethyl 5-(4-t-butylphenyl)-5-oxovalerate

  • MDL number MFCD02261395
  • Cas Number : 101577-33-7
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7650C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7650C
Availability details for
Quantity
  • Availability
  • Add to Cart