RIE156330479 Rieke Metals, Inc.

ethyl 6-(4-t-butylphenyl)-6-oxohexanoate

  • MDL number MFCD02261396
  • Cas Number : 898778-44-4
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7650D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7650D
Availability details for
Quantity
  • Availability
  • Add to Cart