RIE156330481 Rieke Metals, Inc.

Ethyl 8-(4-t-butylphenyl)-8-oxooctanoate

  • MDL number MFCD02261398
  • Cas Number : 898778-50-2
  • Molecular Weight : 318.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7650F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7650F
Availability details for
Quantity
  • Availability
  • Add to Cart