RIE156330485 Rieke Metals, Inc.

ethyl 7-oxo-7-(4-n-pentylphenyl)heptanoate

  • MDL number MFCD02261402
  • Cas Number : 898757-24-9
  • Molecular Weight : 318.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7651E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7651E
Availability details for
Quantity
  • Availability
  • Add to Cart