RIE156330487 Rieke Metals, Inc.

ethyl 5-(4-hexylphenyl)-5-oxovalerate

  • MDL number MFCD02261404
  • Cas Number : 138247-14-0
  • Molecular Weight : 304.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7652C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7652C
Availability details for
Quantity
  • Availability
  • Add to Cart