RIE156330492 Rieke Metals, Inc.

ethyl 5-(2-ethoxyphenyl)-5-oxovalerate

  • MDL number MFCD02261414
  • Cas Number : 898757-39-6
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7654C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7654C
Availability details for
Quantity
  • Availability
  • Add to Cart